Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Latent TGF-beta 1 Protein
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS and Glycerol.
Description
The Recombinant Human Latent TGF-beta 1 Protein is derived from CHO. The Recombinant Human Latent TGF-beta 1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS and Glycerol. |
| Gene ID (Entrez) | 7040 |
| Molecular Weight (g/mol) | Observed Molecular Wt: 40-43kDa & 12kDa, reducing conditions 80-90kDa, non-reducing conditions, Predicted Molecular Wt.: 41.3kDa (monomer) |
| Name | Latent TGF-beta 1 |
| Quantity | 5 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Leu30-Ser390 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, ≤ -20° C under sterile conditions after opening. |
| Termination | Leu30 & Ala279 |
| Endotoxin Concentration | <1.0 EU per 1μg of the protein by the LAL method. |
| Gene Alias | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction