Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Latent TGF-beta 1 Protein

Catalog No. 299LT025CF
Change view
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS and Glycerol.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
299LT025CF 25 μg Supplied as a 0.2μm filtered solution in PBS and Glycerol.
299LT005 5 μg Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein.
299LT005CF 5 μg Supplied as a 0.2μm filtered solution in PBS and Glycerol.
299LT025 25 μg Supplied as a 0.2μm filtered solution in PBS and Glycerol with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 299LT025CF Supplier R&D Systems™ Supplier No. 299LT025/CF
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Latent TGF-beta 1 Protein is derived from CHO. The Recombinant Human Latent TGF-beta 1 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS and Glycerol.
Gene ID (Entrez) 7040
Molecular Weight (g/mol) Observed Molecular Wt: 40-43kDa & 12kDa, reducing conditions 80-90kDa, non-reducing conditions, Predicted Molecular Wt.: 41.3kDa (monomer)
Name Latent TGF-beta 1
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Leu30-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening. 3 months, ≤ -20° C under sterile conditions after opening.
Termination Leu30 & Ala279
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.