Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Latrophilin 3 Protein Fc Chimera
SDP

Catalog No. 8915LN050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8915LN050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8915LN050 Supplier R&D Systems™ Supplier No. 8915LN050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Latrophilin 3 Protein Fc Chimera is derived from CHO. The Recombinant Human Latrophilin 3 Protein Fc Chimera has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_056051.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 23284
Molecular Weight (g/mol) Theoretical M.W.: 120kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Latrophilin 3/LPHN3 protein, (N-terminus) Human Latrophilin 3 (Phe20-Leu854) Accession &Num; NP_056051 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CIRL3, latrophilin 3, latrophilin homolog 3 (cow), LEC3, Lectomedin-3, LPHN3
Common Name Latrophilin 3/LPHN3
Gene Symbol ADGRL3
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human Latrophilin 3/LPHN3 Fc Chimera is immobilized at 0.5μg/mL (100μL/well), the concentration of Recombinant Human FLRT3 that produces 50% of the optimal binding response is approximately 0.07-0.35μg/mL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Human Latrophilin 3 (Phe20-Leu854) Accession # NP_056051, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.