Learn More
R&D Systems™ Recombinant Human Lefty-A Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 7044 |
| Molecular Weight (g/mol) | 39 kDa |
| Name | Lefty-A |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human Lefty-A protein, (N-terminus) Mouse Lefty-1 Propeptide (Leu22-Arg77) Accession &Num; Q64280 HHHHHH Human Lefty-A (Phe78-366) Accession &Num; O00292 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 0.5mg/mL in sterile PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.