Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LEKTI/SPINK5 Protein

Catalog No. 8564PI010
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8564PI010 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8564PI010 Supplier R&D Systems™ Supplier No. 8564PI010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LEKTI/SPINK5 Protein is derived from HEK293. The Recombinant Human LEKTI/SPINK5 Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number Q9NQ38
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 11005
Molecular Weight (g/mol) Observed M.W.: 57-65kDa, reducing conditions, Theoretical M.W.: 51kDa
Quantity 10 μg
Source Human embryonic kidney cell,HEK293-derived human LEKTI/SPINK5 protein Glu626-Glu1064,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias Kazal type 5, LETKI, NETS, serine peptidase inhibitor, Kazal type 5, SPINK5, VAKTI
Common Name SPINK5
Gene Symbol SPINK5
Biological Activity Measured by its ability to inhibit Recombinant Human Kallikrein 7 cleavage of a fluorogenic peptide substrate Mca-RPKPVE-Nval-WRK(Dnp)-NH2. The IC50 value is <70 nM, as measured under the described conditions.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Glu626-Glu1064, with a C-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.