Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Lgr6 Fc Chimera Protein

Catalog No. 8470GP050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8470GP050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8470GP050 Supplier R&D Systems™ Supplier No. 8470GP050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Lgr6 Fc Chimera Protein is derived from CHO. The Recombinant Human Lgr6 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9HBX8
For Use With (Application) Bioactivity
Gene ID (Entrez) 59352
Molecular Weight (g/mol) Observed M.W.: 90-108kDa, reducing conditions, Theoretical M.W.: 87kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Lgr6 protein, (N-terminus) Human Lgr6 (Ala25-Ala567) Accession &Num; Q9HBX8 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631
Common Name Lgr6
Gene Symbol LGR6
Biological Activity Measured by its ability to induce Topflash reporter activity in HEK293T human embryonic kidney cells. The ED50 for this effect is typically 2.5-12.5μg/mL in the presence of 10ng/mL Recombinant Mouse Wnt-3a.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Human Lgr6 (Ala25-Ala567) Accession # Q9HBX8, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330) C-terminus
Purity or Quality Grade >80%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.