Learn More
R&D Systems™ Recombinant Human LH alpha/beta Heterodimer Protein
Description
Specifications
Specifications
| Accession Number | P01215 ( alpha chain) & P01229 ( beta chain) |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | Observed M.W.: 15-26kDa, reducing conditions, Theoretical M.W.: 10kDa ( alpha chain), 13kDa ( beta chain) |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human LH alpha/beta Heterodimer protein Ala25-Ser116 ( alpha chain) & Ser21-Leu141 ( beta chain) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Common Name | LH alpha/beta Heterodimer |
| Biological Activity | Measured by its ability to induce cAMP accumulation in MLTC-1 mouse Leydig tumor cells. Rebois, R.V. et al. (1982) J. Cell Biol. 94:70. The ED50 for this effect is typically 0.25-2.5ng/mL. |
| Conjugate | Unconjugated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.