Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LIF, Biotinylated Protein
SDP

Catalog No. BT7734025CF Shop All Novus Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS
Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
BT7734025CF Lyophilized from a 0.2μm filtered solution in PBS
BT7734025 Lyophilized from a 0.2μm filtered solution in PBS with BSA as carrier protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BT7734025CF Supplier R&D Systems™ Supplier No. BT7734025/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LIF, Biotinylated Protein is derived from E. coli. The Recombinant Human LIF, Biotinylated Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P15018
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 3976
Molecular Weight (g/mol) 20 kDa
Quantity 25 μg
Source E. coli-derived human LIF protein Pro24-Phe202
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Biotinylated protein via amines
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDA, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Biological Activity 0.02 to 0.12ng/mL
Product Type Recombinant protein
Conjugate Biotin
Species Human
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.