Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LILRA6/CD85b/ILT8 Protein
SDP

Catalog No. 9088T4050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9088T4050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 9088T4050 Supplier R&D Systems™ Supplier No. 9088T4050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LILRA6/CD85b/ILT8 Protein is derived from HEK293. The Recombinant Human LILRA6/CD85b/ILT8 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q6PI73
For Use With (Application) Bioactivity
Gene ID (Entrez) 79168
Molecular Weight (g/mol) Observed M.W.: 62-69kDa, reducing conditions, Theoretical M.W.: 47kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human LILRA6/CD85b protein Gly24-Asn447,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Common Name ILT8/CD85b/LILRA6
Gene Symbol LILRA6
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human LILRA6/CD85b/ILT8 is coated onto a microplate at 2μg/mL, Recombinant Human Angiopoietin-like Protein 7/ANGPTL7 binds with a typical ED50 = 0.1-0.6μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Gly24-Asn447, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.