Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LILRB1/CD85j/ILT2 Fc Chimera Protein

Catalog No. 2017T2050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2017T2050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 2017T2050 Supplier R&D Systems™ Supplier No. 2017T2050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to support the adhesion of HSB2 human peripheral blood acute lymphoblastic leukemia cells. Immobilized Recombinant Human LILRB1/CD85j/ILT2 Fc Chimera at 5 μg/mL, 100 μL/well can support 60-80% HSB2 cell adhesion, when 1 x 105 cells were added in each well of a 96-well plate.

Specifications

Accession Number Q8NHL6
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 10859
Molecular Weight (g/mol) M.W. (Observed): 106 kDa, reducing conditions; M.W. (theoretical): 73.8 kDa (monomer)
Name ILT2/CD85j Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human LILRB1/CD85j/ILT2 protein Human LILRB1/CD85j/ILT2 (Gly24-His458) (N-terminus) Accession # Q8NHL6 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol LILRB1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein ILT2/CD85j/LILRB1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.