Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LILRB4/CD85k/ILT3 Fc Chimera Protein

Catalog No. 10429T4050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-429-T4050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-429-T4050 Supplier R&D Systems™ Supplier No. 10429T4050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

Measured by its binding ability in a functional ELISA. When Recombinant Human Apolipoprotein E3/ApoE3 (Catalog # 4144-AE ) is immobilized at 5 μg/mL, 100 μL/well, the concentration of Recombinant Human LILRB4/CD85k/ILT3 Fc Chimera (Catalog # 10429-T4) that produces 50% of the optimal binding response is approximately 0.8-5 μg/mL.

Specifications

Accession Number NP_001265355.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 11006
Molecular Weight (g/mol) M.W. (Observed): 55-75 kDa, under reducing conditions; M.W. (theoretical): 53 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human LILRB4/CD85k/ILT3 protein Human LILRB4/CD85k/ILT3 (Gly24-Glu259) (N-terminus) Accession # NP_001265355.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein ILT3/CD85k/LILRB4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.