Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LILRB5/CD85c/LIR-8 Protein

Catalog No. 8487T4025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-87T-4025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-87T-4025 Supplier R&D Systems™ Supplier No. 8487T4025
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag

Measured by its binding ability in a functional ELISA. When Recombinant Human LILRB5/CD85c/LIR-8 is coated at 2 μg/mL, Recombinant Human Angiopoietin-like Protein 7/ANGPTL7 (Catalog # 914-AN ) binds with an apparent Kd <4 nM.

Specifications

Accession Number O75023
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 10990
Molecular Weight (g/mol) M.W. (Observed): 64-77 kDa, reducing conditions; M.W. (theoretical): 49 kDa
Name LILRB5/CD85c/LIR-8
Preparation Method Reconstitute at 100μg/mL in PBS.
Quality Control Testing SDS-PAGE: 64-77kDa, reducing conditions
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived human LILRB5/CD85c/LIR-8 protein Arg18-His456, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Arg18
Without Additives Carrier Protein
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol LILRB5
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE with silver staining.
Protein LIR-8/CD85c/LILRB5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.