Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human LINGO-1 His Protein
SDP

Catalog No. NBP25201605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25201605 0.5 mg
NBP25201601 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25201605 Supplier Novus Biologicals™ Supplier No. NBP2520160.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 241-337 of Human LINGO-1 The Recombinant Human LINGO-1 Protein is derived from E. coli. The Recombinant Human LINGO-1 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 10% glycerol
Gene ID (Entrez) 84894
Molecular Weight (g/mol) TMW: 15.1kDa
Quantity 0.5 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias FLJ14594, LERN1UNQ201, leucine rich repeat and Ig domain containing 1, leucine rich repeat neuronal 6A, Leucine-rich repeat and immunoglobilin domain-containing protein 1, Leucine-rich repeat neuronal protein 1, Leucine-rich repeat neuronal protein 6A, LRRN6Aleucine-rich repeat neuronal 6A, MGC17422
Common Name LINGO-1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.