Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Lipoprotein Lipase/LPL Protein

Catalog No. 9888LL100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-88L-L100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-88L-L100 Supplier R&D Systems™ Supplier No. 9888LL100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Lipoprotein Lipase/LPL Protein is derived from CHO. The Recombinant Human Lipoprotein Lipase/LPL Protein has been validated for the following applications: Enzyme Activity.

Specifications

Accession Number P06858
Gene ID (Entrez) 4023
Molecular Weight (g/mol) 51 kDa
Name EC 3.1.1, HDLCQ11, LIPD, LIPDEC 3.1.1.34, lipoprotein lipase, LPL
Quantity 100 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Lipoprotein Lipase/LPL protein Ala28-Gly475 with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU per 1 µg of the protein by the LAL method.
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.