Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LRIG1 Protein
SDP

Catalog No. 8504LR050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-04L-R050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 85-04L-R050 Supplier R&D Systems™ Supplier No. 8504LR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LRIG1 Protein is derived from HEK293. The Recombinant Human LRIG1 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q96JA1
For Use With (Application) Bioactivity
Gene ID (Entrez) 26018
Molecular Weight (g/mol) Observed M.W.: 91-120kDa, reducing conditions, Theoretical M.W.: 84kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human LRIG1 protein, (N-terminus) Human LRIG1 (Ala35-Ser779) Accession &Num; Q96JA1 HP GGGSGGGSGGGS HHHHHH (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias D6Bwg0781e, Img, LIG1, LIG-1
Common Name LRIG1
Gene Symbol LRIG1
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of BCE C/D-1b bovine corneal endothelial cells. The ED50 for this effect is typically 0.5-2.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Human LRIG1 (Ala35-Ser779) Accession # Q96JA1, N-terminus; HP; GGGSGGGSGGGS; HHHHHH, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.