Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human LRP-4 Protein

Catalog No. 5948LR025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-48L-R025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 59-48L-R025 Supplier R&D Systems™ Supplier No. 5948LR025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human LRP-4 Protein is derived from CHO. The Recombinant Human LRP-4 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAI36669
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 4038
Molecular Weight (g/mol) 191kDa
Name LRP-4
Preparation Method Reconstitute at 300μg/mL in PBS.
Quality Control Testing SDS-PAGE: 180-225kDa, reducing conditions
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human LRP-4 protein, (N-terminus) Human LRP-4 (Ser21-Ser1725) Accession &Num; AAI36669 GGGSGGGSGGGS 6-His (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ser21 & Gly212
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 80%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.