Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Ly-6G6F Fc Chimera Protein

Catalog No. 10513L6050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10513L6050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10513L6050 Supplier R&D Systems™ Supplier No. 10513L6050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its ability to inhibit anti-CD3 antibody induced IL-2 or IFN-gamma secretion by human T cells. The ED50 for this effect 1-10 μg/mL.

Specifications

Accession Number Q5SQ64.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 259215
Molecular Weight (g/mol) M.W.-observed: 55-62 kDa, under reducing conditions, M.W.-theoretical: 50 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Ly-6G6F protein Human Ly-6G6F (Ala17-Trp235) (N-terminus) Accession # Q5SQ64.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias C6orf21, G6f, locus G6D, LY6G6D, LY6G6F, lymphocyte antigen 6 complex locus protein G6f, lymphocyte antigen 6 complex, locus G6F, NG32
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Ly-6G6F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.