Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human M-CSF Protein
SDP

Catalog No. 216MC010CF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-6MC0-10CF 10 μg
21-6MC1-00CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-6MC0-10CF Supplier R&D Systems™ Supplier No. 216MC010/CF
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells. Nakoinz, I. et al. (1990) J. Immunol. 145:860. The ED50 for this effect is 0.5-1.5 ng/mL.

Specifications

Accession Number NP_757350
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5 kDa (monomer)
Quantity 10 μg
Source <EM>E. coli</EM>-derived human M-CSF protein Glu33-Ser190, with an N-terminal Met
Storage Requirements Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1 μg of the protein by the LAL method.
Gene Symbol CSF1
Conjugate Unconjugated
Cross Reactivity M-CSF
Species Human
Reconstitution Reconstitute at 50-500 μg/mL in sterile PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.