Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human M-CSF Protein

Catalog No. 216MC01M
Change view
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
21-6MC-01M 1 mg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
216MC005 5 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
216MC005CF 5 μg Lyophilized from a 0.2μm filtered solution in PBS.
216MC025 25 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
216MC025CF 25 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 21-6MC-01M Supplier R&D Systems™ Supplier No. 216MC01M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells. Nakoinz, I. et al. (1990) J. Immunol. 145:860. The ED50 for this effect is 0.5-1.5 ng/mL.

Specifications

Accession Number NP_757350
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5kDa (monomer)
Quantity 1 mg
Source E. coli-derived human M-CSF protein Glu33-Ser190,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0EU per 1μg of the protein by the LAL method.
Gene Alias colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim,mgC31930
Gene Symbol CSF1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 50 to 500μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.