Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human M-CSF Protein
GSA_VASDP

Catalog No. 216MC025 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
216MC025 25 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
216MC005 5 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
216MC005CF 5 μg Lyophilized from a 0.2μm filtered solution in PBS.
216MC025CF 25 μg Lyophilized from a 0.2μm filtered solution in PBS.
21-6MC-01M 1 mg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 216MC025 Supplier R&D Systems™ Supplier No. 216MC025

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured in a cell proliferation assay using M-NFS-60 mouse myelogenous leukemia lymphoblast cells. Nakoinz, I. et al. (1990) J. Immunol. 145:860. The ED50 for this effect is 0.5-1.5 ng/mL.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 1435
Molecular Weight (g/mol) 18.5 kDa (monomer)
Name M-CSF
Quantity 25 μg
Source E. coli-derived human M-CSF protein Glu33-Ser190,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 50-500μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Product Line R&D Systems
Purity or Quality Grade 97%, by SDS-PAGE with silver staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.