Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MAD2L1 His Protein
SDP

Catalog No. NBP17245810 Shop All R&D Systems Products
Change view
Click to view available options
:
10μg; Unlabeled
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP17245810 10μg; Unlabeled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP17245810 Supplier Novus Biologicals™ Supplier No. NBP17245810UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-205 of Human MAD2L1 The Recombinant Human MAD2L1 Protein is derived from E. coli. The Recombinant Human MAD2L1 Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number NP_002349
For Use With (Application) SDS-PAGE
Formulation Liquid. 20mM Tris-HCl buffer (pH8.0) containing 20% glycerol, 0.1M NaCl, 1mM DTT
Gene ID (Entrez) 4085
Molecular Weight (g/mol) 25.7kDa
Name MAD2L1 Protein
Purification Method SDS-PAGE
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Cross Reactivity Human
Purity or Quality Grade >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.