Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MAT2A His-tag Protein

Catalog No. 10315MA050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-315-MA050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-315-MA050 Supplier R&D Systems™ Supplier No. 10315MA050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured via hydrolysis of ATP during synthesis of S-adenosylmethionine. The specific activity is >85 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P31153-1
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 4144
Molecular Weight (g/mol) 44 kDa
Name AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2, SAMS2
Quantity 50 μg
Source E. coli-derived human MAT2A protein Asn2-Tyr395 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
Conjugate Unconjugated
Species Human
Buffer Supplied as a 0.2 μm filtered solution in Tris, NaCl, TCEP and Glycerol.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.