Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human MAT2A Protein
SDP

Catalog No. NBC11851505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC11851505 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC11851505 Supplier Novus Biologicals™ Supplier No. NBC1185150.5MG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-395 of Human MAT2A The Recombinant Human MAT2A Protein is derived from E. coli. The Recombinant Human MAT2A Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1.0 mg/ml
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl pH 8.0, 10% glycerol
Gene ID (Entrez) 4144
Molecular Weight (g/mol) 45.8 kDa
Quantity 0.5 mg
Source E. coli
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Symbol MAT2A
Cross Reactivity MAT2A
Species Human
Purity or Quality Grade >95% pure by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.