Learn More
Novus Biologicals™ Recombinant Human Methionine Aminopeptidase 1/METAP1 Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Concentration | 1mg/mL |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 3mM DTT, 40% glycerol, 200mM NaCl, 0.1mM PMSF |
| Gene ID (Entrez) | 23173 |
| Quantity | 100μg |
| Storage Requirements | Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO - research use only |
| Species | Human |
| Recombinant | Recombinant |
| Purity or Quality Grade | >90%, by SDS-PAGE |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.