Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Methionine Aminopeptidase 1D Protein

Catalog No. 4714ZN020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4714ZN020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 4714ZN020 Supplier R&D Systems™ Supplier No. 4714ZN020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC (Catalog # ES017 ). The resulting GP-AMC is cleaved by Recombinant Human DPPIV/CD26 (Catalog # 9168-SE ). The specific activity is >30 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number Q6UB28
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol.
Gene ID (Entrez) 254042
Molecular Weight (g/mol) M.W. (Observed): 38 kDa, reducing conditions; M.W. (theoretical): 34 kDa
Name Methionine Aminopeptidase 1D/MAP1D
Quantity 20 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Methionine Aminopeptidase 1 D/MAP1D protein Arg44-Ala335, with an N-terminal Met and a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol METAP1D
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Methionine Aminopeptidase 1 D/MAP1D
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.