Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human mGluR8 Protein

Catalog No. 9534UR100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-34U-R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 95-34U-R100 Supplier R&D Systems™ Supplier No. 9534UR100
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant HumanmgluR8 Protein CF are coupled to effector systems through GTP-binding proteins and modulate glutamate neurotransmission in the central and peripheral nervous systems.

The Recombinant Human mGluR8 Protein is derived from CHO.

Specifications

Accession Number O00222-1
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 2917
Molecular Weight (g/mol) 55kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line,CHO-derived human mGluR8 protein Gln34-Ser514,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias FLJ40498, glutamate receptor, metabotropic 7, GPRC1G, GRM7, metabotropic glutamate receptor 7,mglu7,mgluR7,mgLUR7GPRC1GGLUR7
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.