Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MGMT His Protein
SDP

Catalog No. NBC118534 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118534 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118534 Supplier R&D Systems™ Supplier No. NBC118534

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-207 of Human MGMT The Recombinant Human MGMT Protein is derived from E. coli. The Recombinant Human MGMT Protein has been validated for the following applications: SDS-Page.

Specifications

Accession Number B4DEE8
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 7.5), 1mM DTT, 10% glycerol
Gene ID (Entrez) 4255
Molecular Weight (g/mol) 23.8kDa
Name MGMT Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen MGMT, 1-207aa, Human, His-tagged, Recombinant, E.coli (NP_002403).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line Novus
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.