Learn More
R&D Systems™ Recombinant Human MIA Protein
Description
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Source | E. coli-derived human MIA protein Gly25-Gln131,with an N-terminal Met |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.