Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MICL/CLEC12A Fc Chimera Protein
SDP

Catalog No. 10835ML050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10835ML050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10835ML050 Supplier R&D Systems™ Supplier No. 10835ML050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human Integrin alpha X beta 2 Protein (Catalog # 5755-AX ) is immobilized at 5.00 μg/mL, 100 μL/well, the concentration of Recombinant Human MICL/CLEC12A Fc Chimera (Catalog# 10835-ML) that produces 50% of the optimal binding response is approximately 1.20-12.0 μg/mL.

Specifications

Accession Number AAI26292.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 160364
Molecular Weight (g/mol) M.W.-observed: 70-85 kDa, under reducing conditions, M.W.-theroretical: 50 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human MICL/CLEC12A protein MD Human IgG1 (Pro100-Lys330) IEGR Human MICL/CLEC12A (Thr67-Ala265) (N-terminus) Accession # AAI26292.1 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein MICL/CLEC12A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.