Learn More
R&D Systems™ Recombinant Human Midkine Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 4192 |
| Molecular Weight (g/mol) | 13.3 kDa |
| Name | Midkine |
| Quantity | 10 μg |
| Source | E. coli-derived human Midkine protein Lys23-Asp143 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Reconstitution | Reconstitute at 100 μg/mL in sterile PBS. Reconstituted rhMK should be used immediately or aliquoted and stored at -20° C to -70° C. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.