Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MIF Protein, CF

Catalog No. 289MF002CF
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
289MF002CF 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 289MF002CF Supplier R&D Systems™ Supplier No. 289MF002/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Applications: Bioactivity

The Recombinant Human MIF Protein is derived from E. coli. The Recombinant Human MIF Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number AAA36315
For Use With (Application) Bioactivity
Gene ID (Entrez) 4282
Molecular Weight (g/mol) 12.4 kDa & 12.3 kDa
Quantity 2 μg
Source E. coli-derived human MIF protein Pro2-Ala115,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 3 months, -20 to -70 °C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity MIF
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer Supplied as a 0.2 μm filtered solution in MES and NaCl.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.