Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MINDY2/FAM63B Protein, CF

Catalog No. E620050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E6-200-50 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. E6-200-50 Supplier R&D Systems™ Supplier No. E620050
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Recombinant Human MINDY-2 is a Ubiquitin-specific deconjugating enzyme that cleaves K48-linked poly-Ubiquitin chains.

Specifications

Accession Number Q8NBR6
For Use With (Application) Bioactivity
Formulation X mg/mL (X μM) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP
Gene ID (Entrez) 54629
Molecular Weight (g/mol) M.W. (theoretical): 67 kDa
Quantity 50 μg
Source E. coli-derived human MINDY2 protein
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 24 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Specific Reactivity MINDY2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Buffer X mg/mL (X μm) in 50 mM HEPES pH 7.5, 100 mM NaCl, 10% (v/v) Glycerol, 1 mM TCEP
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein MINDY2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.