Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human MMP-12 Protein
SDP

Catalog No. 917MPB020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-7MP-B020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-7MP-B020 Supplier R&D Systems™ Supplier No. 917MPB020
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (Catalog # ES001 ). The specific activity is >500 pmol/min/μg, as measured under the described conditions.

Order Info

Ships at ambient temperature.

Specifications

Accession Number P39900
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in MES, NaCl, CaCl2, CHAPS, ZnSO4 and PEG.
Gene ID (Entrez) 4321
Molecular Weight (g/mol) M.W. (Observed): 53-60 kDa, reducing conditions; M.W. (theoretical): 52 kDa
Quantity 20 μg
Source Mouse myeloma cell line, NS0-derived human MMP-12 protein Leu17-Cys470, with Val97Leu substitution
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 3.4.24, hME, HMEEC 3.4.24.65
Biological Activity >500pmol/min/μg
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 0.25 mg/mL in 10 mM MES, 0.1 M NaCl, 100 μM CaCl2, 0.1% CHAPS, 1 μM ZnSO4 and 0.1% PEG., pH 6.0.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein MMP-12
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.