Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat Activin A, ACFP Protein

Catalog No. ACFP338
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACFP338 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. ACFP338 Supplier R&D Systems™ Supplier No. ACFP338
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. Schwall, R.H. et al. (1991) Method Enzymol. 198:340. The ED50 for this effect is 0.2-1.2 ng/mL. The specific activity of Recombinant Human/Mouse/Rat Activin A is approximately 1 IU/μg, which is calibrated against human Activin A WHO International Standard (NIBSC code: 91/626).

Specifications

Accession Number P08476
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HCl.
Gene ID (Entrez) 3624
Molecular Weight (g/mol) M.W. (Observed): 13 kDa, reducing conditions; M.W. (theoretical): 13 kDa (monomer)
Quantity 50 μg
Source Spodoptera frugiperda, Sf 9 (baculovirus)-derived Activin A protein Met1-Ser426 Produced in an animal component free process (ACFP).
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol INHBA
Specific Reactivity Activin A
Conjugate Unconjugated
Species Human, Mouse, Rat
Recombinant Recombinant
Reconstitution Reconstitute at 100-500 μg/mL in sterile 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in HCl.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Activin A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.