Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed)
SDP

Catalog No. 355BEC100CF Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-5BE-C100CF 100 μg
35-5BE-C025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-5BE-C100CF Supplier R&D Systems™ Supplier No. 355BEC100/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. ED50 for this effect is 0.08 to 0.48 μg/mL.

Specifications

Accession Number P12643
For Use With (Application) Bioactivity
Gene ID (Entrez) 650
Molecular Weight (g/mol) 12.8 kDa (monomer)
Name BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Quantity 100 μg
Source E. coli-derived BMP-2 protein Ala284-Arg396 This form is not detected using human BMP-2 Quantikine ELISA (Catalog # DBP200 ).
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol BMP2
Conjugate Unconjugated
Species Human, Mouse, Rat
Reconstitution Reconstitute at 100 μg/mL in sterile 4 mM HCl.
Buffer Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA.
Endotoxin Level <0.01 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.