Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 Protein
SDP

Catalog No. p-4719153 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
355BM010CF 10 μg Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
355BM010 10 μg Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid with BSA as a carrier protein.
355BM050 50 μg Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid with BSA as a carrier protein.
355BM050CF 50 μg Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 355BM010CF Supplier R&D Systems™ Supplier No. 355BM010/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Binnerts, M.E. et al. (2004) Biochem. Biophys. Res. Commun. 315(2):272. The ED50 for this effect is 40-200 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Glycine, Sucrose, Tween™ 80 and Glutamic Acid.
Gene ID (Entrez) 650
Molecular Weight (g/mol) 13 kDa (monomer)
Name BMP-2
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived BMP-2 protein Gln283-Arg396
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.