Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat Irisin Protein

Catalog No. 8880IR025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8880IR025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8880IR025 Supplier R&D Systems™ Supplier No. 8880IR025
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human/Mouse/Rat Irisin Protein is derived from CHO. The Recombinant Human/Mouse/Rat Irisin Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q8NAU1
For Use With (Application) Bioactivity
Gene ID (Entrez) 252995
Molecular Weight (g/mol) Observed M.W.: 15-37kDa, reducing conditions, Theoretical M.W.: 13kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived Irisin/FNDC5 protein Asp32-Glu143,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias fibronectin type III domain containing 5, fibronectin type III domain-containing protein 5, FNDC5, FRCP2, FRCP2Fibronectin type III repeat-containing protein 2
Common Name Irisin/FNDC5
Gene Symbol FNDC5
Biological Activity Measured by its ability to induce p38 MAPK activation in 3T3-L1 mouse embryonic fibroblast adipose-like cells. 1μg/mL of Recombinant Human Irisin can effectively induce p38 MAPK activation.
Conjugate Unconjugated
Species Human, Mouse, Rat
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Asp32-Glu143, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.