Learn More
Description
Specifications
Specifications
| Accession Number | Q15843.1 |
| Formulation | Supplied as a 0.2 μm filtered solution in HEPES, NaCl and DTT. |
| Gene ID (Entrez) | 4738 |
| Molecular Weight (g/mol) | 8.6 kDa |
| Name | NEDD8 |
| Quantity | 500 μg |
| Source | E. coli-derived NEDD8 protein Met1 - Gly76 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
