Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Measured by its ability to activate MSP R/Ron in MDA-MB-453 or T47D human breast cancer cells. 80 ng/mL of the recombinant human MSP significantly induces phosphorylation of MSP R/Ron measured by DuoSet IC human phospho-MSP R/Ron kit (Catalog # DYC1947 ). Measured by its binding ability in a functional ELISA. When Recombinant Human MSPR/Ron Protein (Catalog #1947-MS) is represent at 100 ng/mL (100 μg/mL), The concentration of Recombinant Human MSP/MST1 that produces 50% optimal binding response is found to be 8-40 ng/mL.
Specifications
Specifications
| Accession Number | AAA59872 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 4485 |
| Molecular Weight (g/mol) | M.W. (Observed): 81 kDa, 62 kDa and 30 kDa, reducing conditions; M.W. (theoretical): 78.5 kDa (MSP alpha/beta); 53.3 kDa (alpha chain), 25.2 kDa (beta chain) |
| Name | MSP/MST1 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human MSP/MST1 protein Gln19-Gly711 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
