Learn More
R&D Systems™ Recombinant Human MSP/MST1 Protein
Description
Specifications
Specifications
| Accession Number | AAA59872 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 4485 |
| Molecular Weight (g/mol) | M.W. (Observed): 81 kDa, 62 kDa and 30 kDa, reducing conditions; M.W. (theoretical): 78.5 kDa (MSP alpha/beta); 53.3 kDa (alpha chain), 25.2 kDa (beta chain) |
| Name | MSP/MST1 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived human MSP/MST1 protein Gln19-Gly711 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked heterodimer |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.