Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human N-Acetylglucosaminyltransferase1/MGAT1
SDP

Catalog No. 8334GT025 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-34G-T025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-34G-T025 Supplier R&D Systems™ Supplier No. 8334GT025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to transfer N-Acetyl-D-Glucosamine from UDP-GlcNAc to alpha 1-3, alpha 1-6-Mannotriose. The specific activity is >30 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number P26572
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 4245
Molecular Weight (g/mol) M.W. (Observed): 45-52 kDa, reducing conditions; M.W. (theoretical): 48 kDa
Name N-Acetylglucosaminyltransferase I/MGAT1
Quality Control Testing SDS-PAGE: 45-52kDa, reducing conditions
Quantity 25 μg
Source Mouse myeloma cell line, NS0-derived human N-Acetylglucosaminyltransferase I/MGAT1 protein Thr30-Asn445, with C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Regulatory Status RUO
Termination Thr30
Without Additives Carrier Protein
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol MGAT1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane.)
Protein MGAT1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.