Learn More
R&D Systems™ Recombinant Human N-Acetylmannosamine Kinase/GNE Protein
Description
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately
Specifications
Specifications
| Accession Number | Q9Y223 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and DTT. |
| Gene ID (Entrez) | 10020 |
| Molecular Weight (g/mol) | M.W. (Observed): 31-39 kDa, reducing conditions; M.W. (theoretical): 34 kDa |
| Name | N-Acetylmannosamine Kinase/GNE, Carrier Free |
| Quantity | 50 μg |
| Source | E. coli-derived human N-Acetylmannosamine Kinase/GNE protein Thr406-Arg720, with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.