Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human NCAM-1/CD56 Fc Chimera Protein

Catalog No. 10136NC050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-136-NC050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-136-NC050 Supplier R&D Systems™ Supplier No. 10136NC050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Neural cell adhesion molecule 1 (NCAM-1), also known as CD56, is a multifunctional member of the Ig superfamily

  • It belongs to a family of membrane-bound glycoproteins that are involved in Ca++ independent cell matrix and homophilic or heterophilic cell-cell interactions.
  • NCAM-1 in the adult brain shows a decline of sialylation relative to earlier developmental periods.
  • In regions that retain a high degree of neuronal plasticity, however, the adult brain continues to express polysialylated-NCAM-1, suggesting sialylation of NCAM-1 is involved in regenerative processes and synaptic plasticity
  • NCAM can function as a receptor for GDNF family of ligands. GDNF stimulates Schwann cell migration and neurite outgrowth in hippocampal and cortical neurons through binding to NCAM-1 and activation of Fyn, independently of RET
  • NCAM-1 also interacts with Ig IIIC isoform of the FGFR1 and plays an important role in developmental events as well as tumor progression
  • Due to expression on immune cells NCAM-1 has great implications on tumor progression, infection, and disease development

Specifications

Accession Number P13591-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 4684
Molecular Weight (g/mol) 103 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human NCAM-1/CD56 protein

Human NCAM-1/CD56 (Leu20-Gly708) Accession # P13591-1IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 2 weeks, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.