Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 4684 |
| Molecular Weight (g/mol) | 105 kDa, reducing conditions |
| Name | NCAM-1/CD56 120 isoform |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human NCAM-1/CD56 protein, (N-terminus) Human NCAM-1 (Leu20-Pro603) Accession &Num; NP_001070150 R Human NCAM-1 (Glu636-Asn741) Accession &Num; NP_001070150 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | CD56, CD56 NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
