Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Nectin-2/CD112 Fc Chimera Protein

Catalog No. 9317N2050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93-17N-2050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 93-17N-2050 Supplier R&D Systems™ Supplier No. 9317N2050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human Nectin-2/CD112 Fc Chimera is immobilized at 0.5 μg/mL (100 μL/well), the concentration of Recombinant Human DNAM-1 Fc Chimera (Catalog # 666-DN ) that produces 50% of the optimal binding response is 0.04-0.2 μg/mL.

Order Info

Ships at ambient temperature

Specifications

Accession Number NP_002847.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5819
Molecular Weight (g/mol) M.W. (theoretical): 62 kDa
Name Nectin-2/CD112 Fc Chimera Recombinant Protein
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Nectin-2/CD112 protein Human Nectin-2/CD112 (Gln32-Leu360) (C-terminus) Accession # NP_002847.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Common Name CD112 antigen; CD112; Herpes virus entry mediator B; Herpesvirus entry mediator B; herpesvirus entry protein B; HVEB; HVEBpoliovirus receptor-like 2; MPH; nectin 2; Nectin2; Nectin-2; poliovirus receptor-related 2 (herpesvirus entry mediator B); polioviru
Biological Activity 0.04 to 0.2μg/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE with silver staining, under reducing conditions.
Protein Nectin-2/CD112
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.