Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Nectin-2/CD112 Protein

Catalog No. 2229N2050CF
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in PBS.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
2229N2050CF Supplied as a 0.2 μm filtered solution in PBS.
2229N2050 Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 2229N2050CF Supplier R&D Systems™ Supplier No. 2229N2050/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to bind with Recombinant Human DNAM-1/CD226 Fc Chimera (Catalog # 666-DN ) in a functional ELISA.

Specifications

Accession Number NP_002847
For Use With (Application) Binding Activity
Formulation Supplied as a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5819
Molecular Weight (g/mol) M.W. (Observed): 43-50 kDa, reducing conditions; M.W. (theoretical): 36.3 kDa
Name Nectin-2/CD112
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human Nectin-2/CD112 protein Gln32-Leu360, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening.
Structural Form Monomer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol NECTIN2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution It is recommended that sterile PBS be added to the vial to prepare a working stock solution of no less than 50 μg/mL The carrier-free protein should be used immediately upon reconstitution to avoid losses in activity due to non-specific binding to the inside surface of the vial. For long term storage as a dilute solution, a carrier protein (e.g. 0.1% HSA or BSA) should be added to the vial.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Protein Nectin-2/CD112
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.