Learn More
R&D Systems™ Recombinant Human NELL2 Protein
Description
Specifications
Specifications
| Accession Number | Q99435 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 4753 |
| Molecular Weight (g/mol) | Observed M.W.: 100-125kDa, reducing conditions, Theoretical M.W.: 90kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human NELL2 protein Leu22-Leu816,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Gene Alias | Mel91, nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2 |
| Common Name | NELL2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.