Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and EDTA. |
| Gene ID (Entrez) | 9423 |
| Molecular Weight (g/mol) | 67.5 kDa |
| Name | Netrin-1 |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human Netrin-1 protein, (N-terminus) Human Netrin-1 (Val22-Ala604) Accession &Num; O95631 HPGGGSGGGSGGGS 6-His tag (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Product Line | R&D Systems |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
