Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Neuregulin-1 beta 1/NRG1 beta 1 Animal-Free Protein
SDP

Catalog No. N276272100U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N276272100U 100 μg
N276272500U 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N276272100U Supplier Novus Biologicals™ Supplier No. NBP276272100UG

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page

An un-tagged recombinant protein corresponding to Human Neuregulin-1 beta 1/NRG1 beta 1

The Recombinant Human Neuregulin-1 beta 1/NRG1 beta 1 Animal-Free Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration Lyoph
For Use With (Application) SDS-PAGE
Gene ID (Entrez) 3084
Molecular Weight (g/mol) 7.6 kDa
Quantity 100 μg
Source E. coli
Storage Requirements Store at -20 to -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration Acceptance criteria for endotoxin levels are <1 EUs/ug protein (determined by Kinetic LAL Method).
Gene Symbol NRG1
Specific Reactivity Neuregulin-1 beta 1/NRG1 beta 1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Sterile water at 0.1 mg/mL
Buffer Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA)
Purity or Quality Grade >95%, by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.