Learn More
R&D Systems™ Recombinant Human Neuroligin 1v2/NLGN1v2 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 22871 |
| Molecular Weight (g/mol) | 71.3 kDa |
| Name | Neuroligin 1/NLGN1 Variant 2 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived human Neuroligin 1/NLGN1 protein Met1-Ser677,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.